missing translation for 'onlineSavingsMsg'
Learn More

TLX/NR2E1 Antibody (SR1840), Novus Biologicals™

Product Code. 18676234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18676234 100 μg 100µL
18699364 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18676234 Supplier Novus Biologicals Supplier No. NBP322225100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TLX/NR2E1 Monoclonal antibody specifically detects TLX/NR2E1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX/NR2E1
Applications Western Blot
Classification Monoclonal
Clone SR1840
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias hTll, nuclear receptor subfamily 2, group E, member 1, Nuclear receptor TLX, Protein tailless homolog, tailes-related receptor, tailless homolog, Tll, TLXnuclear receptor subfamily 2 group E member 1, XTLL
Host Species Rabbit
Immunogen A synthesized peptide derived from human TLX/NR2E1 (Uniprot #: Q9Y466)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7101
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.