Learn More
TLR3 Antibody (27N3D4), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-27405PCP
This item is not returnable.
View return policy
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| TLR3 | |
| Monoclonal | |
| PerCP | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 27N3D4 | |
| Flow Cytometry 1 μL/1 million cells | |
| TLR3 | |
| A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7098 | |
| Store at 4C in the dark. | |
| IgG2b κ |