Learn More
TLR3 Antibody (27N3D4), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-27405PCP
Product Code. 18171174
  • 611.00€ / 0.10mL
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLR3
Monoclonal
PerCP
CD283, CD283 antigen, toll-like receptor 3
Rat
Protein G purified
RUO
Primary
Human, Mouse
Purified
Flow Cytometry
27N3D4
Flow Cytometry 1 μL/1 million cells
TLR3
A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
0.1 mL
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
7098
Store at 4C in the dark.
IgG2b κ