missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (CL10552) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21176G
This item is not returnable.
View return policy
Description
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western Blot
Specifications
| TLR2 | |
| Monoclonal | |
| DyLight 488 | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| This antibody was developed using a synthetic peptide derived from O60603, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 7097 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL10552 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction