missing translation for 'onlineSavingsMsg'
Learn More

TLL1 Mouse anti-Human, Clone:4H8C1, Abnova™

Product Code. 16044926
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16044926 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16044926 Supplier Abnova Supplier No. MAB17642.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant human TLL1.

This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. A similar protein in mice is required during heart development and specifically processes procollagen C-propeptides and chordin at similar cleavage sites. [provided by RefSeq]

Specifications

Antigen TLL1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4H8C1
Conjugate Unconjugated
Description tolloid-like 1
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene TLL1
Gene Alias TLL
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 870-1013 of human TLL1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 7092
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.