missing translation for 'onlineSavingsMsg'
Learn More

TLK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen TLK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18263483
View Documents
Novus Biologicals
NBP1-54355
100 μL
483.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TLK2 Polyclonal specifically detects TLK2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TLK2
Polyclonal
Rabbit
Cell Cycle and Replication
EC 2.7.11, EC 2.7.11.1, MGC44450, PKU-ALPHA, serine/threonine-protein kinase tousled-like 2, tousled-like kinase 2serine/threonine kinase
TLK2
IgG
82 kDa
Western Blot
Unconjugated
RUO
Q86UE8
11011
Synthetic peptides corresponding to TLK2(tousled-like kinase 2) The peptide sequence was selected from the N terminal of TLK2. Peptide sequence VSAQQNSPSSTGSGNTEHSCSSQKQISIQHRQTQSDLTIEKISALENSKN.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.