missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLE4 Monoclonal Antibody (384CT24.1.3.4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537554
This item is not returnable.
View return policy
Description
TLE4 Monoclonal Antibody for Western Blot
Transcriptional corepressor that binds to a number of transcription factors. Inhibits the transcriptional activation mediated by PAX5, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES.
Specifications
| TLE4 | |
| Monoclonal | |
| 1.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q04727 | |
| TLE4 | |
| KLH conjugated synthetic peptide between 262-290 amino acids from the Central region of human TLE4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Western Blot | |
| 384CT24.1.3.4 | |
| Unconjugated | |
| TLE4 | |
| 5730411M05Rik; AA792082; B lymphocyte gene 1; BCE1; BCE-1; E(spI); enhancer of split groucho 4; ESG; ESG4; Esp2; ESTM13; ESTM14; GRG4; Grg-4; groucho-related protein 4; KIAA1261; Protein ESP2; TLE family member 4, transcriptional corepressor; Tle4; transducin like enhancer of split 4; transducin-like enhancer of split 4; transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4, E(spl) homolog; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); transducin-like enhancer of split 4, homolog of Drosophila E(spl); transducin-like enhancer protein 4; X83332; X83333 | |
| Mouse | |
| Euglobulin Precipitation | |
| RUO | |
| 7091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction