Learn More
Macherey-Nagel™ TLC Simultaneous Developing Chamber
Product Code. 12374613 Shop All Macherey-Nagel Products
Change view
Click to view available options
Holds:
2
5
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
12374613 2 1 set
10486471 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12374613 Supplier Macherey-Nagel™ Supplier No. 814018
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 13-10-2026
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
  • Accessory for TLC
  • Chambers are thick walled
  • Clear glass tank 
  • Heavy ground glass lid
Specifications
Quantity 1 Pk.
Type TLC Simultaneous Developing Chamber
For Use With (Application) For 2 Plates 10 x 10 cm
Holds 2