Learn More
Macherey-Nagel™ TLC Micro Kit for Beginners
Product Code. 12460087 Shop All Macherey-Nagel Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
12460087 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12460087 Supplier Macherey-Nagel™ Supplier No. 814000
  • ONLINE EXCLUSIVE PRICE
    350.00€
    296.95€ / Pack of 1
    Save 53.05€
    15% Off
 Restricted Item
In Stock
TLC introductory kit for beginners, science education and method development
TLC Micro-Set A for beginners.
This Micro-Set A contains all chemicals and accessories for the following experiments:
  • Separation of a mixture of fat-soluble / lipophilic dyes (test dye mixture 1) butter yellow, indophenol, sudan blue II, sudan red G
  • Separation of a mixture of anthraquinone dyes (test dye mixture 2) blue 1, blue 3, green, green blue, red, violet 1, violet 2
  • Separation of a mixture of food dyes (test dye mixture 3) brilliant black BN (E151), fast red E, erythrosine (E127), yellow orange S (sunset yellow CFC, E110), naphthol red S, ponceau 4 R (E124), tartrazine (E102)
  • Separation of dyes from felt tip pens.
This product contain harmful substances which must be specially labeled as hazardous. For detailed information please see SDS.
Specifications
For Use With (Application) Science Education
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • TLC Micro Set A
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: ammonia solution, 2-propanol, ethanol, toluene, cyclohexane