Learn More
Applied Biosystems™ TaqMan™ Universal Extraction Control Organism
Shop All Applied Biosystems ProductsDescription
The TaqMan Universal Extraction Control Organism control is used with the proprietary TaqMan assay for Bacillus atrophaeus (Assay ID Ba06596576_s1). Like other gram-positive bacteria, Bacillus atrophaeus has thick cell walls than can be difficult to lyse. This characteristic makes gram-positive bacteria an ideal control to monitor the efficiency of cell lysis and subsequent nucleic acid recovery.
TaqMan Universal Extraction Control Organism is supplied lyophilized in a quantity of 1 × 109 copies/vial and is reconstituted in 200 μL of 1X PBS, pH 7.4 to a final concentration 5 × 16 copies/μL. During nucleic acid isolation, 10 μL of the control is processed as a stand-alone sample in a background of universal transport media. It can be added to the negative extraction control, and may also be added to one or more test samples at the start of the extraction process. The control is carried through the remainder of the workflow with the test samples. It is recommended that at least one stand-alone control sample is run per extraction plate.
Specifications
Specifications
| Concentration | 1 x 109 Copies/Vial |
| Content And Storage | 3 vials containing lyophilized Bacillus atrophaeus. Store at 4°C. |
| Detection Method | Primer-probe |
| Format | Vial |
| Product Line | TaqMan |
| Quantity | 3 pellets |
| Shipping Condition | Room temperature |
| For Use With (Application) | Pathogen Detection |
| No. of Reactions | 60 Reactions |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.