missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titin Antibody (T11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
215.00€
Specifications
| Antigen | Titin |
|---|---|
| Clone | T11 |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen 1:10-1:500, Electron Microscopy 1:10-1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Titin Monoclonal specifically detects Titin in Human, Mouse, Rat, Avian, Chicken, Fish, Insect, Mammal, Reptile samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Electron Microscopy.Specifications
| Titin | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen 1:10-1:500, Electron Microscopy 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| cardiomyopathy, dilated 1G (autosomal dominant), CMD1G, CMH9, CMPD4, Connectin, DKFZp451N061, EC 2.7.11.1, EOMFC, FLJ32040, FLJ34413, FLJ39564, FLJ43066, HMERF, LGMD2JFLJ26020, MPRM, Rhabdomyosarcoma antigen MU-RMS-40.14, titin, TMDFLJ26409 | |
| TTN | |
| IgG2b | |
| Unpurified | |
| The antibody localizes titin (connectin) in skeletal and heart muscle of a wide variety of species from cold-blooded vertebrates to human. The antibody does not cross-react with nebulin. Likewise, it does not react with smooth muscle, non-muscle tissues, or cultured cells. |
| T11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers | |
| Ascites with 0.1% Sodium Azide | |
| 7273 | |
| Full length native protein (purified) (Chicken breast muscle). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title