Learn More
Titanium Standard Metal Solution 1000ppm, for AAS, Solution HCl 2M, Fisher Chemical™
Product Code. p-7073226 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 10611193
Brand: Fisher Chemical J/8110/05
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Titanium Standard Metal Solution 1000ppm, for AAS, Solution HCl 2M
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 11264
Specifications
Packaging HDPE plastic bottle
Physical Form Liquid
Color Purple
Quantity 100 mL