Learn More
Titanium, plasma standard solution, Specpure™ Ti 1000μg/mL
Product Code. 11378997
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Product Code. 11378997
Brand: Thermo Scientific Alfa Aesar 038720.AD
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number MFCD00011264
Specifications
Physical Form Liquid
Quantity 50 mL
Grade Specpure
Chemical Name or Material Titanium plasma standard solution, Ti 1000μg/mL