missing translation for 'onlineSavingsMsg'
Learn More

Tirofiban Hydrochloride Hydrate, TRC

Product Code. 30137725
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137725 250 mg 250mg
30008437 50 mg 50mg
17971565 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137725 Supplier Toronto Research Chemicals Supplier No. TRCT446750250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150915-40-5
Molecular Formula C22 H36 N2 O5 S . Cl H . H2 O
Molecular Weight (g/mol) 495.07
Synonym N-(Butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-L-tyrosine hydrochloride hydrate (1:1:1), L-Tyrosine, N-(butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-, monohydrochloride, monohydrate (9CI), Tirofiban hydrochloride monohydrate
IUPAC Name (2S)-2-(butylsulfonylamino)-3-[4-(4-piperidin-4-ylbutoxy)phenyl]propanoic acid;hydrate;hydrochloride
SMILES O.Cl.CCCCS(=O)(=O)N[C@@H](Cc1ccc(OCCCCC2CCNCC2)cc1)C(=O)O

Specifications

InChI Formula InChI=1S/C22H36N2O5S.ClH.H2O/c1-2-3-16-30(27,28)24-21(22(25)26)17-19-7-9-20(10-8-19)29-15-5-4-6-18-11-13-23-14-12-18;;/h7-10,18,21,23-24H,2-6,11-17H2,1H3,(H,25,26);1H;1H2/t21-;;/m0../s1
Quantity 250 mg
Formula Weight 494.2217
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Tirofiban, Hydrochloride Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.