CAS RN 150915-40-5
CAS RN 150915-40-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tirofiban Hydrochloride Hydrate, TRC
CAS: 150915-40-5 Molecular Formula: C22 H36 N2 O5 S . Cl H . H2 O Molecular Weight (g/mol): 495.07 Synonym: N-(Butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-L-tyrosine hydrochloride hydrate (1:1:1),L-Tyrosine, N-(butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-, monohydrochloride, monohydrate (9CI),Tirofiban hydrochloride monohydrate IUPAC Name: (2S)-2-(butylsulfonylamino)-3-[4-(4-piperidin-4-ylbutoxy)phenyl]propanoic acid;hydrate;hydrochloride SMILES: O.Cl.CCCCS(=O)(=O)N[C@@H](Cc1ccc(OCCCCC2CCNCC2)cc1)C(=O)O