Learn More
Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
Product Code. 18708203 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18708203 - -
18467731 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18708203 Supplier Novus Biologicals™ Supplier No. NBP226245
  • ONLINE EXCLUSIVE PRICE
    705.00€
    666.75€ / 1mg
    Save 38.25€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Applications: Flow Cytometry, In vitro assay
TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da