missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Mouse anti-Human, Clone: TIMP2/2044, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07720-100ug
This item is not returnable.
View return policy
Description
TIMP-2 Monoclonal specifically detects TIMP-2 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| TIMP-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| Recombinant human TIMP-2 protein fragment (aa 48-220) (exact sequence is proprietary) | |
| 100 μg | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 7077 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| TIMP2/2044 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction