missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIF1 alpha Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33244-20ul
This item is not returnable.
View return policy
Description
TIF1 alpha Monoclonal antibody specifically detects TIF1 alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TIF1 alpha | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human TIF1 alpha (O15164).,, Sequence:, TNNTIQFHCDPSFWAQNIINLGSLVIEDKESQPQMPKQNPVVEQNSQPPSGLSSNQLSKFPTQISLAQLRLQHMQQQVMAQRQQVQRRPAPVGLPNPRMQG | |
| 20 μL | |
| Protein Kinase, Zinc Finger | |
| 8805 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction