missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticlopidine N-Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Ticlopidine N-Oxide
Specifications
Specifications
| Chemical Name or Material | Ticlopidine N-Oxide |
| CAS | 79923-55-0 |
| Analyte or Component Names | Ticlopidine N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 Cl N O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Thieno[3,2-c]pyridine, 5-[(2-chlorophenyl)methyl]-4,5,6,7-tetrahydro-, 5-oxide, Ticlopidine N-Oxide |
| InChI Formula | InChI=1S/C14H14ClNOS/c15-13-4-2-1-3-11(13)9-16(17)7-5-14-12(10-16)6-8-18-14/h1-4,6,8H,5,7,9-10H2 |
| SMILES | [O-][N+]1(Cc2ccccc2Cl)CCc3sccc3C1 |
| IUPAC Name | 5-[(2-chlorophenyl)methyl]-5-oxido-6,7-dihydro-4H-thieno[3,2-c]pyridin-5-ium |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction