missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TICAM2 Monoclonal Antibody (757706)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561424
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
TRAM, or TRIF-related adaptor molecule (TICAM2), plays an essential role in the MyD88-independent signaling of TLR4. The functional absence of TRAM in vascular endothelial cells restricts TLR4 signaling to MyD88-dependent pathway. Overexpression of TRAM cDNA in endothelial cells reconstituted LPS-induced Trif-dependent NF-kB activation and cxcl10 promoter activity.
Specifications
| TICAM2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8BJQ4 | |
| TICAM2 | |
| E. coli-derived recombinant mouse TRAM/TICAM2, Met1-Ala232. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 757706 | |
| Unconjugated | |
| TICAM2 | |
| B430113A10; cytoplasmic adaptor; MyD88-4; NF-kappa-B-activating protein 502; putative NF-kappa-B-activating protein 502; TAG; TICAM2; TICAM-2; TIR domain-containing adapter molecule 2; TIR domain-containing adapter protein; TIRAP3; Tirp; toll like receptor adaptor molecule 2; toll/interleukin-1 receptor (TIR) domain-containing adapter protein; toll/interleukin-1 receptor domain-containing protein; Toll-interleukin I receptor domain (TIR)-containing adaptor molecule (TICAM) 2; toll-like receptor adaptor molecule 2; toll-like receptor adaptor protein 3; Tram; Trif; TRIF-related adapter molecule; TRIF-related adapter molecule TRAM; TRIF-related adaptor molecule | |
| Rat | |
| Protein A/G | |
| RUO | |
| 225471 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction