missing translation for 'onlineSavingsMsg'
Learn More

Tiapride Hydrochloride, TRC

Product Code. 30135354
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135354 25 g 25g
30132973 100 g 100g
17924135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135354 Supplier Toronto Research Chemicals Supplier No. TRCT43705025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51012-33-0
Molecular Formula C15 H24 N2 O4 S . Cl H
Molecular Weight (g/mol) 364.89
Synonym N-[2-(Diethylamino)ethyl]-2-methoxy-5-(methylsulfonyl)-benzamide monohydrochloride, Gramalil, Italprid, Luxoben, Sereprile, Tiapridal, Tiapride hydrochloride, Tiapridex
IUPAC Name N-[2-(diethylamino)ethyl]-2-methoxy-5-methylsulfonylbenzamide;hydrochloride
SMILES Cl.CCN(CC)CCNC(=O)c1cc(ccc1OC)S(=O)(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H24N2O4S.ClH/c1-5-17(6-2)10-9-16-15(18)13-11-12(22(4,19)20)7-8-14(13)21-3;/h7-8,11H,5-6,9-10H2,1-4H3,(H,16,18);1H
Formula Weight 364.1224
Percent Purity >95%
Chemical Name or Material Tiapride hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.