accessibility menu, dialog, popup

UserName

CAS RN 51012-33-0

HClH3CNH3CNHOOCH3SH3COO

CAS RN 51012-33-0

IUPAC Name: hydrogen N-[2-(diethylamino)ethyl]-5-methanesulfonyl-2-methoxybenzamide chloride
Synonyms: hydrogen tiapride chloride
Molecular Weight (g/mol): 364.89
Molecular Formula: C15H25ClN2O4S
InChi Key: OTFDPNXIVHBTKW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCNC(=O)C1=CC(=CC=C1OC)S(C)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results