missing translation for 'onlineSavingsMsg'
Learn More

Tiagabine Hydrochloride, Mikromol™

Product Code. 30051118
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30051118

Brand: Mikromol MM1029.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tiagabine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tiagabine Hydrochloride
CAS 145821-59-6
Analyte or Component Names Tiagabine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H25 N O2 S2 . Cl H
Certifications/Compliance ISO 17025
Synonym Tiagabine Hydrochloride, (3R)-1-[4,4-Bis(3-methyl-2-thienyl)-3-butenyl]-3-piperidinecarboxylic acid hydrochloride, (R)-1-[4,4-Bis(3-methyl-2-thienyl)-3-butenyl]-3-piperidinecarboxylic acid hydrochloride, ABT 569, Abbott 70569 hydrochloride, Abbott 70569.1, Gabitril, NNC 05-0328, NO 05-0328, Tiabex, Tiagabine hydrochloride
InChI Formula InChI=1S/C20H25NO2S2.ClH/c1-14-7-11-24-18(14)17(19-15(2)8-12-25-19)6-4-10-21-9-3-5-16(13-21)20(22)23;/h6-8,11-12,16H,3-5,9-10,13H2,1-2H3,(H,22,23);1H/t16-;/m1./s1
SMILES Cl.Cc1ccsc1C(=CCCN2CCC[C@H](C2)C(=O)O)c3sccc3C
IUPAC Name (3R)-1-[4,4-bis(3-methylthiophen-2-yl)but-3-enyl]piperidine-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 412.01
Quantity 250 mg
Formula Weight 411.1093 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.