missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine 4’-O-Beta-D-Glucuronide, TRC

Product Code. 17942825
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942825 10 mg 10mg
17984785 5 mg 5mg
17973935 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942825 Supplier Toronto Research Chemicals Supplier No. TRCT42563010MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21462-56-6
Molecular Formula C21H19I4NO10
Molecular Weight (g/mol) 952.99
Synonym 4-[4-[(2S)-2-Amino-2-carboxyethyl]-2,6-diiodophenoxy]-2,6-diiodophenyl β-D-Glucopyranosiduronic ccAcid, Levothyroxine Phenolic Glucuronide, (S)-4-[4-(2-Amino-2-carboxyethyl)-2,6-diiodophenoxy]-2,6-diiodophenyl β-D-Glucopyranosiduronic Acid, L-Thyroxine Glucuronide, Thyroxine Glucuronide;
IUPAC Name (2S,3S,4S,5R,6S)-6-[4-[4-[(2S)-2-amino-2-carboxyethyl]-2,6-diiodophenoxy]-2,6-diiodophenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O[C@H]3[C@@H]([C@H]([C@@H]([C@H](O3)C(=O)O)O)O)O)I)I)C[C@@H](C(=O)O)N

Specifications

InChI Formula InChI=1S/C21H19I4NO10/c22-8-1-6(3-12(26)19(30)31)2-9(23)16(8)34-7-4-10(24)17(11(25)5-7)35-21-15(29)13(27)14(28)18(36-21)20(32)33/h1-2,4-5,12-15,18,21,27-29H,3,26H2,(H,30,31)(H,32,33)/t12-,13-,14-,15+,18-,21+/m0/s1
Quantity 10 mg
Formula Weight 952.7188
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Thyroxine 4'-O-beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.