CAS RN 21462-56-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thyroxine 4’-O-Beta-D-Glucuronide, TRC
CAS: 21462-56-6 Molecular Formula: C21H19I4NO10 Molecular Weight (g/mol): 952.99 Synonym: 4-[4-[(2S)-2-Amino-2-carboxyethyl]-2,6-diiodophenoxy]-2,6-diiodophenyl β-D-Glucopyranosiduronic ccAcid,Levothyroxine Phenolic Glucuronide,(S)-4-[4-(2-Amino-2-carboxyethyl)-2,6-diiodophenoxy]-2,6-diiodophenyl β-D-Glucopyranosiduronic Acid,L-Thyroxine Glucuronide,Thyroxine Glucuronide; IUPAC Name: (2S,3S,4S,5R,6S)-6-[4-[4-[(2S)-2-amino-2-carboxyethyl]-2,6-diiodophenoxy]-2,6-diiodophenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O[C@H]3[C@@H]([C@H]([C@@H]([C@H](O3)C(=O)O)O)O)O)I)I)C[C@@H](C(=O)O)N