Learn More
Thyroxine 4’-O-Beta-D-Glucuronide, TRC
Product Code. 17984785
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984785 5 mg 5mg
17942825 10 mg 10mg
17973935 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17984785 Supplier Toronto Research Chemicals Supplier No. TRCT4256305MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21462-56-6
Molecular Formula C21H19I4NO10
Molecular Weight (g/mol) 952.99
Synonym 4-[4-[(2S)-2-Amino-2-carboxyethyl]-2,6-diiodophenoxy]-2,6-diiodophenyl β-D-Glucopyranosiduronic ccAcid, Levothyroxine Phenolic Glucuronide, (S)-4-[4-(2-Amino-2-carboxyethyl)-2,6-diiodophenoxy]-2,6-diiodophenyl β-D-Glucopyranosiduronic Acid, L-Thyroxine Glucuronide, Thyroxine Glucuronide;
IUPAC Name (2S,3S,4S,5R,6S)-6-[4-[4-[(2S)-2-amino-2-carboxyethyl]-2,6-diiodophenoxy]-2,6-diiodophenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O[C@H]3[C@@H]([C@H]([C@@H]([C@H](O3)C(=O)O)O)O)O)I)I)C[C@@H](C(=O)O)N
Specifications
InChI Formula InChI=1S/C21H19I4NO10/c22-8-1-6(3-12(26)19(30)31)2-9(23)16(8)34-7-4-10(24)17(11(25)5-7)35-21-15(29)13(27)14(28)18(36-21)20(32)33/h1-2,4-5,12-15,18,21,27-29H,3,26H2,(H,30,31)(H,32,33)/t12-,13-,14-,15+,18-,21+/m0/s1
Quantity 5 mg
Formula Weight 952.7188
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Thyroxine 4'-O-beta-D-Glucuronide
Grade HPLC
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.