missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid Peroxidase Rabbit anti-Human, Clone: TPO/3813R, Novus Biologicals™
Rabbit Monoclonal Antibody
528.00€
Specifications
| Antigen | Thyroid Peroxidase |
|---|---|
| Clone | TPO/3813R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Thyroid Peroxidase | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 7173 | |
| IgG | |
| Protein A or G purified |
| TPO/3813R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| Recombinant fragment of human Thyroid Peroxidase (around aa 685-804) (Exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title