Learn More
Thymidine glycol Antibody (B29), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-11041C
This item is not returnable.
View return policy
Description
Thymidine glycol Monoclonal antibody specifically detects Thymidine glycol in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| Thymidine glycol | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| B29 | |
| ELISA, Immunohistochemistry | |
| 32645-65-1, 5,6-dihydro-5,6-dihydroxythymidine, 5,6-dihydroxy-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,3-diazinane-2,4-dione, 5,6-Dihydroxy-5,6-dihydrothymidine, THD glycol, Thymidine glycol, Thymidine, 5,6-dihydro-5,6-dihydroxy- | |
| Thymidine glycol polymer | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |