missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THSD7A Antibody (CL3779) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61639G
This item is not returnable.
View return policy
Description
THSD7A Monoclonal antibody specifically detects THSD7A in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| THSD7A | |
| Monoclonal | |
| DyLight 488 | |
| thrombospondin, type I, domain containing 7A | |
| This antibody was developed using a recombinant protein derived from Q9UPZ6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology | |
| 221981 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3779 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction