Learn More
Thiencarbazone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Thiencarbazone |
| CAS | 936331-72-5 |
| Analyte or Component Names | Thiencarbazone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H12 N4 O7 S2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 4-[[[(4,5-Dihydro-3-methoxy-4-methyl-5-oxo-1H-1,2,4-triazol-1-yl)carbonyl]amino]sulfonyl]-5-methyl-3-thiophenecarboxylic acid, Thiencarbazone |
| InChI Formula | InChI=1S/C11H12N4O7S2/c1-5-7(6(4-23-5)8(16)17)24(20,21)13-9(18)15-11(19)14(2)10(12-15)22-3/h4H,1-3H3,(H,13,18)(H,16,17) |
| Solution Type | Neat |
| SMILES | COC1=NN(C(=O)NS(=O)(=O)c2c(C)scc2C(=O)O)C(=O)N1C |
| Show More |