missing translation for 'onlineSavingsMsg'
Learn More

Thiamethoxam-desmethyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325502
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325502 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325502 Supplier Dr. Ehrenstorfer Supplier No. DREA17453013AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamethoxam-desmethyl
CAS 171103-04-1
Analyte or Component Names Thiamethoxam-desmethyl
Shipping Condition Room Temperature
Molecular Formula C7 H8 Cl N5 O3 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2H-1,3,5-Oxadiazin-4-amine, 3-[(2-chloro-5-thiazolyl)methyl]-3,6-dihydro-N-nitro-, 3-[(2-Chloro-5-thiazolyl)methyl]-3,6-dihydro-N-nitro-2H-1,3,5-oxadiazin-4-amine, CGA 330050, Desmethylthiamethoxam, N-Desmethylthiamethoxam
InChI Formula InChI=1S/C7H8ClN5O3S/c8-6-9-1-5(17-6)2-12-4-16-3-10-7(12)11-13(14)15/h1H,2-4H2,(H,10,11)
SMILES [O-][N+](=O)NC1=NCOCN1Cc2cnc(Cl)s2
IUPAC Name N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-2,6-dihydro-1,3,5-oxadiazin-4-yl]nitramide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 277.6881
Quantity 1 mL
Formula Weight 277.0036 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.