missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ThermoPrime 2x ReddyMix PCR Master Mix, with 1.5 mM MgCl2
Description
Thermo Scientific ThermoPrime Taq DNA Polymerase with 10X ReddyMix PCR Buffer. This unique buffer has an inert red dye and a density reagent added. After thermal cycling, a sample of the PCR mix may be removed and loaded directly onto an agarose gel without the addition of loading dye. ReddyMix has no adverse effect on the outcome of PCR; yields are the same as with standard ThermoPrime Taq DNA Polymerase. Convenient Master Mix format allows easier reaction set-up.
Features
Enhanced enzyme stability – for increased yield
Saves time – no need to add gel loading dye
Advanced buffering system – minimizes PCR optimization steps
Convenient Master Mix formats – save time and reduce the risk of sample contamination during assay setup
Applications
Amplifying fragments up to 5 kb
Routine PCR
Related Products
ThermoPrime Taq DNA Polymerase, with 10x ReddyMix PCR buffer and separate vial of 25 mM MgCl2
ThermoPrime 2x ReddyMix PCR Master Mix, with 1.5 mM MgCl2
Features
Applications
Related Products
ThermoPrime Taq DNA Polymerase, with 10x ReddyMix PCR buffer and separate vial of 25 mM MgCl2
ThermoPrime 2x ReddyMix PCR Master Mix, with 1.5 mM MgCl2
Specifications
Specifications
| Content And Storage | 2 x 1mL ReddyMix PCR Master Mix with 1.5mM MgCl2 (2X). Store at –20°C |
| Description | ThermoPrime 2x ReddyMix PCR Master Mix, 160 rxns, With 1.5mM MgCl2, 1 X Fidelity (vs. Taq), 3'-A Product Overhang, Standard Reaction Speed, –20°C Storage Temperature, Enhanced Enzyme Stability - For Increased Yield, For PCR |
| GC-Rich PCR Performance | Low |
| Polymerase | ThermoPrime taq DNA polymerase |
| Reaction Speed | Standard |
| Product Type | ThermoPrime 2x ReddyMix PCR Master Mix |
| Quantity | 160 rxn(s) |
| For Use With (Application) | Standard PCR |
| Fidelity (vs. Taq) | 1 X |
| No. of Reactions | 160 Reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?