Learn More
Applied Biosystems™ Thermal Tape (for the 310 Genetic Analyzer)
Product Code. 15809706 Shop All Applied Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
310 Genetic Analyzer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15809706 310 Genetic Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15809706 Supplier Applied Biosystems™ Supplier No. 310021
 Restricted Item
In Stock
This is a roll of 0.5 ft. yellow lab marking thermal tape for affixing the capillary to the 310 Genetic Analyzer heat plate.
  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
For Use With (Equipment) 310 Genetic Analyzer
Product Type Tape
Shipping Condition Room Temperature