missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TGF-beta 2 Antibody (TGFB2/1679), Novus Biologicals™

Mouse Monoclonal Antibody

232.00€ - 462.00€

Specifications

Antigen TGF-beta 2
Clone TGFB2/1679
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043476
View Documents
Novus Biologicals™
NBP3-26734-20ug
20 μg
232.00€
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043322
View Documents
Novus Biologicals™
NBP3-26734-100ug
100 μg
462.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TGF-beta 2 Monoclonal antibody specifically detects TGF-beta 2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TGF-beta 2
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Tumor Suppressors
10 mM PBS with 0.05% BSA
7042
IgG1 κ
Protein A or G purified
TGFB2/1679
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2
Recombinant full-length human TGF-beta 2 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.