missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1, 2, 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 143 publications
Brand: R&D Systems MAB1835-100
This item is not returnable.
View return policy
Description
TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Western Blot, Binding Inhibition, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| TGF beta 1/2/3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGFB1 | |
| Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
| 100 μg | |
| Primary | |
| Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. No cross-reactivity was observed with recombinant LAP in direct ELISAs. | |
| Multi-species | |
| Supernatant |
| Western Blot, In vitro Assay, Neutralization, ELISA, Immunocytochemistry | |
| 1D11 | |
| Western Blot 1 μg/mL, Binding Inhibition, Neutralization 0.25-1.25 μg/mL, ELISA Capture (Matched Antibody Pair) 2-8 μg/mL | |
| ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7040 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction