missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR (Transferrin R) Antibody (13E4cc), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
553.00€
Specifications
| Antigen | TfR (Transferrin R) |
|---|---|
| Clone | 13E4cc |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 750 |
Description
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for Western Blot, ELISA, ELISASpecifications
| TfR (Transferrin R) | |
| Western Blot | |
| Alexa Fluor 750 | |
| Mouse | |
| Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 50mM Sodium Borate | |
| 7037 | |
| IgG2a | |
| Protein A purified |
| 13E4cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified human soluble transferrin receptor. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title