missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI2 Monoclonal Antibody (OTI5H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805493
This item is not returnable.
View return policy
Description
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.Specifications
| TFPI2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48307 | |
| TFPI2 | |
| Human recombit protein fragment corresponding to amino acids 23-235 of human TFPI2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H3 | |
| Unconjugated | |
| TFPI2 | |
| PP5, REF1, TFPI-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7980 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction