missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFPI2 Monoclonal Antibody (243220)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561173
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
Specifications
| TFPI2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P48307 | |
| TFPI2 | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI-2, Asp23-Phe235. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 243220 | |
| Unconjugated | |
| TFPI2 | |
| AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7980 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction