missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
202.00€ - 573.00€
Specifications
| Antigen | TFG |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227443
|
Novus Biologicals
NBP3-37919-20ul |
20 μL |
202.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231426
|
Novus Biologicals
NBP3-37919-100ul |
100 μL |
573.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFG Polyclonal antibody specifically detects TFG in Human samples. It is validated for ELISA,Western BlotSpecifications
| TFG | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10342 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human TFG (TFG) (NP_006061.2).,, Sequence:, MNGQLDLSGKLIIKAQLGEDIRRIPIHNEDITYDELVLMMQRVFRGKLLSNDEVTIKYKDEDGDLITIFDSSDLSFAIQCSRILKLTLFVNGQPRPLESSQVKYLRRELIELRNKVNRLLDSLEPPGEPGPSTNIPENDTVDGREEKSASDSSGKQSTQVMAASMSAFDPLKNQDEINKNVMSAFGLTDDQVSGPPSAPAEDRSGTPDSIASSSSAAHPPGVQPQQPPYTGAQTQAGQIE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title