missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody (BLR070G), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77360
This item is not returnable.
View return policy
Description
TFEB Monoclonal antibody specifically detects TFEB in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TFEB | |
| Monoclonal | |
| Unconjugated | |
| Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| BLR070G | |
| Western Blot 1:1000, Immunoprecipitation 20 μg/mg lysate | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| Immunogen was a peptide representing a region between residue 426 and the C-terminus (residue 476) of human Transcription factor EB using the numbering given in entry NP_009093.1 (Gene ID 7942). | |
| 100 μg | |
| Autophagy | |
| 7942 | |
| Store at 2°C to 8°C, 1 year from date of receipt | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction