missing translation for 'onlineSavingsMsg'
Learn More

TFCP2, Mouse anti-Human, Clone: 3H6, Abnova™

Product Code. 16116515
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16116515 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16116515 Supplier Abnova Supplier No. H00007024M01.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence: KHEDGDSNGTFFVYHAIYLEELTAVELTEKIAQLFSISPCQISQIYKQGPTGIHVLISDEMIQNFQEEACFILDTMKAETNDSYHIILK

Specifications

Antigen TFCP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3H6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene transcription factor CP2
Gene Accession No. BC003634
Gene Alias CP2/LBP-1C/LSF/SEF/TFCP2C
Gene Symbols TFCP2
Host Species Mouse
Immunogen TFCP2 (AAH03634, 414 a.a. ∼ 502 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7024
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.