missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144490
This item is not returnable.
View return policy
Description
TFC4 Polyclonal Antibody for Western Blot, ELISA
TFIIIC mediates tRNA and 5S RNA gene activation by binding to intragenic promoter elements. Upstream of the transcription start site, TFIIIC assembles the initiation complex TFIIIB-TFIIIC-tDNA, which is sufficient for RNA polymerase III recruitment and function. Part of the tauA domain of TFIIIC that binds boxA DNA promoter sites of tRNA and similar genes. TFC4 is the TFIIIB-assembling subunit of TFIIIC and essential for viability.
Specifications
| TFC4 | |
| Polyclonal | |
| Unconjugated | |
| TFC4 | |
| PCF1 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 852938 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.755 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P33339 | |
| TFC4 | |
| Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) TFC4 protein. | |
| 100 μL | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction