missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB1M Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35512-100ul
This item is not returnable.
View return policy
Description
TFB1M Polyclonal antibody specifically detects TFB1M in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TFB1M | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| CGI75, CGI-75, dimethyladenosine transferase 1, mitochondrial, EC 2.1.1, EC 2.1.1.-, h-mtTFB, h-mtTFB1, homolog of yeast mitochondrial transcription factor B, hTFB1M, Mitochondrial 12S rRNA dimethylase 1, mitochondrial dimethyladenosine transferase 1, Mitochondrial transcription factor B1, mtTFB, mtTFB1, S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1, transcription factor B1, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-346 of human TFB1M (NP_057104.2).,, Sequence:, QAAKQLSQNFLLDLRLTDKIVRKAGNLTNAYVYEVGPGPGGITRSILNADVAELLVVEKDTRFIPGLQMLSDAAPGKLRIVHGDVLTFKVEKAFSESLKRPWEDDPPNVHIIGNLPFSVSTPLIIKWLENISCRDGPFVYGRTQMTLTFQKEVAERLAANTGSKQRSRLSVMAQYLCNVRHIFTIPGQAFVPKPEVDVGVVHFTPLIQPKIEQPFKLVEKVVQNVFQFRRKYCHRGLRMLFPEAQRLESTGRLLELADIDPTLRPRQLSISHFKSLCDVYRKMCDEDPQLFAYNFREELKRRKSKNEEKEEDDAENYRL | |
| 100 μL | |
| Stem Cell Markers | |
| 51106 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction