missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tetranectin Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148972
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
Specifications
| Tetranectin | |
| Polyclonal | |
| Unconjugated | |
| Clec3b | |
| CLEC3B; C-type lectin domain family 3 member B; C-type lectin domain family 3, member b; LOC100912012; Plasminogen kringle 4-binding protein; tetranectin; tetranectin (plasminogen binding protein); tetranectin (plasminogen-binding protein); tetranectin-like; TN; TNA | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 7123 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P05452 | |
| Clec3b | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin, Glu22-Val202. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction