Learn More
Thermo Scientific Chemicals Tetrabromophenol Blue
Description
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications
Specifications
| Melting Point | 203°C to 205°C (decomposition) |
| Beilstein | 378438 |
| Solubility Information | Soluble in water. |
| Formula Weight | 985.55 |
| Quantity | 5 g |
| Chemical Name or Material | Tetrabromophenol Blue |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.