missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium borohydride, 97%

Product Code. 11412000
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11412000

Brand: Thermo Scientific Alfa Aesar A17494.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33725-74-5 | C16H40BN | 257.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33725-74-5
Molecular Formula C16H40BN
Molecular Weight (g/mol) 257.31
MDL Number MFCD00012035
InChI Key GMBOFJFPOCGSOI-UHFFFAOYSA-N
Synonym tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
PubChem CID 9881569
SMILES [BH4-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 125°C to 131°C
Flash Point 140°C (284°F)
Linear Formula [CH3(CH2)3]4NBH4
Quantity 50 g
UN Number UN3131
Sensitivity Moisture sensitive
IUPAC Name tetrabutylazanium boranuide
Formula Weight 257.31
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium borohydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.