accessibility menu, dialog, popup

UserName

CAS RN 33725-74-5

NH3CH3CCH3CH3BHHHH

CAS RN 33725-74-5

IUPAC Name: tetrabutylazanium boranuide
Synonyms: tetrabutylammonium ion boranuide
Molecular Weight (g/mol): 257.31
Molecular Formula: C16H40BN
InChi Key: GMBOFJFPOCGSOI-UHFFFAOYSA-N
SMILES: [BH4-].CCCC[N+](CCCC)(CCCC)CCCC
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)