missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
202.00€ - 573.00€
Specifications
| Antigen | TET2 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229181
|
Novus Biologicals
NBP3-35271-100ul |
100 μL |
573.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232510
|
Novus Biologicals
NBP3-35271-20ul |
20 μL |
202.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TET2 Polyclonal antibody specifically detects TET2 in Mouse samples. It is validated for ELISA,Western BlotSpecifications
| TET2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1833-2002 of human TET2 (NP_001120680.1).,, Sequence:, VQGVASGAEDNDEVWSDSEQSFLDPDIGGVAVAPTHGSILIECAKRELHATTPLKNPNRNHPTRISLVFYQHKSMNEPKHGLALWEAKMAEKAREKEEECEKYGPDYVPQKSHGKKVKREPAEPHETSEPTYLRFIKSLAERTMSVTTDSTVTTSPYAFTRVTGPYNRYI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 54790 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title