Learn More
Testosterone EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Testosterone EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 3-Ethoxyandrosta-3,5-dien-17-one (Androstenedione Ethylenolether) |
| CAS | 972-46-3 |
| Analyte or Component Names | 3-Ethoxyandrosta-3,5-dien-17-one (Androstenedione Ethylenolether) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C21 H30 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Testosterone Imp. B (EP), 3-Ethoxyandrosta-3,5-dien-17-one, Androstenedione Ethylenolether, Testosterone Impurity B |
| InChI Formula | InChI=1S/C21H30O2/c1-4-23-15-9-11-20(2)14(13-15)5-6-16-17-7-8-19(22)21(17,3)12-10-18(16)20/h5,13,16-18H,4,6-12H2,1-3H3/t16-,17-,18-,20-,21-/m0/s1 |
| SMILES | CCOC1=CC2=CC[C@H]3[C@@H]4CCC(=O)[C@@]4(C)CC[C@@H]3[C@@]2(C)CC1 |
| IUPAC Name | (8R,9S,10R,13S,14S)-3-ethoxy-10,13-dimethyl-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-one |
| Show More |