Learn More
Testosterone Cypionate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Testosterone Cypionate |
| CAS | 58-20-8 |
| Analyte or Component Names | Testosterone Cypionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H40 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Testosterone, cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid, ester with testosterone (8CI), DepAndro 100, DepAndro 200, Depo, Depo-testosterone, Deposteron, Depotest, Depovirin, NSC 9157, Pertestis, Testex Leo, Testodrin prolongatum, Testosterone cyclopentylpropionate, Testosterone cypionate, Testosterone, 3-cyclopentanepropionate, Virilon, Androst-4-en-3-one, 17-(3-cyclopentyl-1-oxopropoxy)-, (17beta)-, 17beta-Hydroxyandrost-4-en-3-one cyclopentanepropionate, 17beta-Hydroxyandrost-4-en-3-one cyclopentylpropionate, Testosterone 17beta-cyclopentanepropionate, Testosterone 17beta-cyclopentylpropionate, Testosterone 17beta-cypionate |
| InChI Formula | InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5 |
| Show More |