missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Antibody (BGN/3G8A4), HRP, Novus Biologicals™

Product Code. 18053724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18053724 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18053724 Supplier Novus Biologicals Supplier No. NB10062943H
  • ONLINE EXCLUSIVE PRICE
    614.00€
    580.65€ / 0.1mL
    Save 33.35€
    5% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Testosterone
Applications ELISA
Classification Monoclonal
Clone BGN/3G8A4
Conjugate HRP
Dilution ELISA
Formulation PBS with No Preservative
Host Species Mouse
Immunogen BSA conjugated testosterone-3,-CMO
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Developmental Biology
Primary or Secondary Primary
Test Specificity This product recognises testosterone, a steroid hormone and most potent of the naturally occurring androgens. Testosterone promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes, but is also produced in small amounts by the adrenal glands, ovaries and placenta. Synthetic derivatives of testosterone are used therapeutically in the treatment of breast cancer and for hormone replacement therapy. NB100-62943 also recognises 5-a-Dihydrotestosterone (1%) and Androstenedione (1%).
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.